missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Xylene-alpha,alpha,alpha,alpha',alpha',alpha'-d6, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | o-Xylene dimethyl-d6 |
| CAS | 25319-54-4 |
| Molecular Formula | C8 2H6 H4 |
| Quantity | 50 mg |
| Synonym | o-Xylene D6 (Dimethyl D6), o-Xylene-α,α,α,α',α',α'-d6 (8CI), 1,2-Di(methyl-d3)benzene, 1,2-Bis(trideuteriomethyl)benzene, o-Xylene-d6, Benzene','1,2-di(methyl-d3)- |
| InChI Formula | InChI=1S/C8H10/c1-7-5-3-4-6-8(7)2/h3-6H,1-2H3/i1D3,2D3 |
| SMILES | [2H]C([2H])([2H])c1ccccc1C([2H])([2H])[2H] |
| IUPAC Name | 1,2-bis(trideuteriomethyl)benzene |
| Molecular Weight (g/mol) | 112.2 |
| Formula Weight | 112.1159 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction