missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NCT 501 hydrochloride

Description
NCT 501 hydrochloride is a potent and selective ALDH1A1 inhibitor (IC50 = 40 nM). Exhibits >1000-fold selectivity for ALDH1A1 over ALDH3A1, ALDH1B1, ALDH2, HPGD and HSD17β4. Also selective over a panel of 168 GPCRs. Blood-brain barrier permeable.
Specifications
Specifications
| Chemical Name or Material | 8-[[4-(Cyclopropylcarbonyl)-1-piperazinyl]methyl]-3,7-dihydro-1,3-dimethyl-7-(3-methylbutyl)-1H-purine-2,6-dione hydrochloride |
| CAS | 1307896-46-3 |
| Quantity | 50 mg |
| Solubility | Soluble to 100mM in water and to 100mM in DMSO |
| Molecular Formula | C21H32N6O3.HCl |
| Formula Weight | 452.98 |
| Physical Form | Solid |
| Color | White |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
For Research Use Only.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?