missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(trans-4-Ethylcyclohexyl)carbonyl]-D-phenylalanine (Nateglinide Ethyl Analog), Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
N-[(trans-4-Ethylcyclohexyl)carbonyl]-D-phenylalanine (Nateglinide Ethyl Analog)
Specifications
Specifications
| Chemical Name or Material | N-[(trans-4-Ethylcyclohexyl)carbonyl]-D-phenylalanine (Nateglinide Ethyl Analog) |
| CAS | 105746-45-0 |
| Analyte or Component Names | N-[(trans-4-Ethylcyclohexyl)carbonyl]-D-phenylalanine (Nateglinide Ethyl Analog) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H25 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | N-[(trans-4-Ethylcyclohexyl)carbonyl]-D-phenylalanine, Nateglinide Imp. E (Pharmeuropa), Nateglinide Ethyl Analog, Nateglinide Impurity E |
| InChI Formula | InChI=1S/C18H25NO3/c1-2-13-8-10-15(11-9-13)17(20)19-16(18(21)22)12-14-6-4-3-5-7-14/h3-7,13,15-16H,2,8-12H2,1H3,(H,19,20)(H,21,22)/t13-,15-,16-/m1/s1 |
| SMILES | CC[C@@H]1CC[C@H](CC1)C(=O)N[C@H](Cc2ccccc2)C(=O)O |
| IUPAC Name | (2R)-2-[(4-ethylcyclohexanecarbonyl)amino]-3-phenylpropanoic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction