missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 315 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ML 315 hydrochloride is a inhibitor of cdc2-like kinases (Clk) and DYRK kinases (IC50 values are 68, 68 and 231 nM for Clk1, Clk4 and Clk2, respectively, and 282 and 1156 nM for Dyrk1A and Dyrk1B). Shows 20-fold selectivity for Clk4 over Dyrk1A. Stable in both mouse and human plasma. Moderately cell permeable.
Specifications
Specifications
| CAS | 1440251-53-5 |
| Quantity | 50 mg |
| Target | Cdc2-like Kinase |
| Molecular Formula | C18H13Cl2N3O2.HCl |
| Formula Weight | 410.68g/mol |
| Product Type | Inhibitor |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction