missing translation for 'onlineSavingsMsg'
Learn More

Melanotan II Acetic Acid Salt, TRC

Product Code. 30003273
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003273 50 mg 50mg
30135639 100 mg 100mg
30011900 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003273 Supplier Toronto Research Chemicals Supplier No. TRCM20875050MG
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C50H69N15O9 .(xCH3COOH)
Molecular Weight (g/mol) 1024.186005
Synonym N-Acetyl-L-norleucyl-L-α-aspartyl-L-histidyl-D-phenylalanyl-L-arginyl-L-tryptophyl-L-lysinamide Cyclic (2→7)-peptide Acetic Acid Salt, 1,4,7,10,13,18-Hexaazacyclotricosane Cyclic Peptide Deriv. Acetic Acid Salt, 17: PN: US20130203670 SEQID: 17 claimed protein, 17: PN: WO2006012667 PAGE: 40 Claimed Protein Acetic Acid Salt, 19: PN: WO2008025074 SEQID: 19 Claimed Protein Acetic Acid Salt, 19: PN: WO2008025094 SEQID: 19 claimed protein, 1: PN: WO2009033712 PAGE: 103 Claimed Protein Acetic Acid Salt, 1: PN: WO2013067309 SEQID: 91 Claimed Protein Acetic Acid Salt, 2: PN: WO2006060873 SEQID: 2 claimed protein Acetic Acid Salt, MT II Acetic Acid Salt, PT 14 Acetic Acid Salt
SMILES O=C([C@H](CCCNC(N)=N)NC([C@H](CC1=CC=CC=C1)NC([C@@H](NC([C@@H](NC([C@H](CCCC)NC(C)=O)=O)CC(NCCCC[C@H](NC2=O)C(N)=O)=O)=O)CC3=CN=CN3)=O)=O)N[C@H]2CC4=CNC5=C4C=CC=C5

Specifications

Quantity 50 mg
Chemical Name or Material Melanotan II Acetic Acid Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.