missing translation for 'onlineSavingsMsg'
Learn More

Ibutamoren Mesylate, MedChemExpress

Product Code. 16434027
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16434027

Brand: Medchem Express HY50844

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ibutamoren Mesylate (MK-677) is a potent, non-peptide Growth hormone secretagogue receptor (GHSR) agonist. Ibutamoren Mesylate is an orally active growth hormone (GH) secretagogue.

Ibutamoren Mesylate (MK-677) is a potent, non-peptide Growth hormone secretagogue receptor (GHSR) agonist. Ibutamoren Mesylate is an orally active growth hormone (GH) secretagogue.

Specifications

Chemical Name or Material Ibutamoren Mesylate
CAS 159752-10-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C28H40N4O8S2
Quantity 50 mg
Synonym MK-677 MK-0677
Solubility Information DMSO : 50 mg/mL (80.03 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (80.03 mM)
SMILES CC(N)(C(N[C@@H](C(N1CCC2(CN(C3=C2C=CC=C3)S(=O)(C)=O)CC1)=O)COCC4=CC=CC=C4)=O)C.CS(=O)(O)=O
Molecular Weight (g/mol) 624.77
Formula Weight 624.77
Percent Purity 97.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.