missing translation for 'onlineSavingsMsg'
Learn More

Granisetron N-Oxide, Mikromol™

Product Code. 30050517
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity unitSize
30050517 50 mg 50mg
1 options
This item is not returnable. View return policy

Product Code. 30050517

Brand: Mikromol MM0398.11

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Granisetron N-Oxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Granisetron N-Oxide
CAS 160177-68-4
Analyte or Component Names Granisetron N-Oxide
Shipping Condition Ice pack (-20°C)
Molecular Formula C18 H24 N4 O2
Certifications/Compliance ISO 9001
Synonym Granisetron N-Oxide, 1H-Indazole-3-carboxamide, 1-methyl-N-(9-methyl-9-oxido-9-azabicyclo[3.3.1]non-3-yl)-, endo- (9CI), 1H-Indazole-3-carboxamide, 1-methyl-N-(9-methyl-9-azabicyclo[3.3.1]non-3-yl)-, N-oxide, endo-
InChI Formula InChI=1S/C18H24N4O2/c1-21-16-9-4-3-8-15(16)17(20-21)18(23)19-12-10-13-6-5-7-14(11-12)22(13,2)24/h3-4,8-9,12-14H,5-7,10-11H2,1-2H3,(H,19,23)/t12-,13+,14-,22?
SMILES Cn1nc(C(=O)N[C@H]2C[C@H]3CCC[C@@H](C2)[N+]3(C)[O-])c4ccccc14
IUPAC Name 1-methyl-N-[(1R,5S)-9-methyl-9-oxido-9-azoniabicyclo[3.3.1]nonan-3-yl]indazole-3-carboxamide
Molecular Weight (g/mol) 328.41
Quantity 50 mg
Formula Weight 328.1899 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.