missing translation for 'onlineSavingsMsg'
Learn More

Gilteritinib, MedChemExpress

Product Code. 16451387
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16451387

Brand: Medchem Express HY12432

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gilteritinib (ASP2215) is a potent and ATP-competitive FLT3/AXL inhibitor with IC50s of 0.29 nM/0.73 nM, respectively.

Gilteritinib (ASP2215) is a potent and ATP-competitive FLT3/AXL inhibitor with IC50s of 0.29 nM/0.73 nM, respectively.

Specifications

Chemical Name or Material Gilteritinib
CAS 1254053-43-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C29H44N8O3
Quantity 50 mg
Synonym ASP2215
Solubility Information Ethanol : 100 mg/mL (180.93 mM; ultrasonic and adjust pH to 2 with HCl) ∣DMSO : 2 mg/mL (3.62 mM; Need ultrasonic)
SMILES NC(C1=NC(CC)=C(NC2CCOCC2)N=C1NC3=CC(OC)=C(N4CCC(N5CCN(C)CC5)CC4)C=C3)=O
Molecular Weight (g/mol) 552.71
Formula Weight 552.71
Percent Purity 97.77%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.