missing translation for 'onlineSavingsMsg'
Learn More

FLT3-IN-2, MedChemExpress

Product Code. 16403637
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16403637 50 mg 50mg
16497526 100 mg 100mg
16447618 10 mg 10mg
16440088 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16403637 Supplier Medchem Express Supplier No. HY18744
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress FLT3-IN-2 is a FLT3 inhibitor with IC50 of < 1 μM, detailed information refer to WO 2012158957 A2 and WO 2007013896.

FLT3-IN-2 is a FLT3 inhibitor with IC50 of < 1 μM, detailed information refer to WO 2012158957 A2 and WO 2007013896.

Specifications

Chemical Name or Material FLT3-IN-2
CAS 923562-23-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H16ClF3N4
Quantity 50 mg
Solubility Information DMSO : ≥ 30 mg/mL (71.97 mM)
SMILES FC(C1=CC=C(CNC2=NC=C(CC3=CNC4=NC=C(Cl)C=C43)C=C2)C=C1)(F)F
Molecular Weight (g/mol) 416.83
Formula Weight 416.83
Percent Purity 98.38%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.