missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Valacyclovir Hydrochloride, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | D-Valaciclovir Hydrochloride |
| CAS | 1795786-28-5 |
| Molecular Formula | C13 H20 N6 O4 . Cl H |
| Quantity | 50 mg |
| Synonym | D-Valine','2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]ethyl ester','hydrochloride (1:1), GW 277421A, D-valacyclovir, Ph Eur Valaciclovir Impurity R |
| InChI Formula | InChI=1S/C13H20N6O4.ClH/c1-7(2)8(14)12(21)23-4-3-22-6-19-5-16-9-10(19)17-13(15)18-11(9)20;/h5,7-8H,3-4,6,14H2,1-2H3,(H3,15,17,18,20);1H/t8-;/m1./s1 |
| SMILES | Cl.CC(C)[C@@H](N)C(=O)OCCOCn1cnc2C(=O)NC(=Nc12)N |
| IUPAC Name | 2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]ethyl (2R)-2-amino-3-methylbutanoate;hydrochloride |
| Molecular Weight (g/mol) | 360.8 |
| Formula Weight | 360.1313 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction