missing translation for 'onlineSavingsMsg'
Learn More

D-erythro-Sphingosine, MedChemExpress

Product Code. 16409796
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16409796

Brand: Medchem Express HY101047

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress D-erythro-Sphingosine (Erythrosphingosine) is a very potent activator of p32-kinase with an EC50 of 8 μM, and inhibits protein kinase C (PKC). D-erythro-Sphingosine (Erythrosphingosine) is also a PP2A activator.

D-erythro-Sphingosine (Erythrosphingosine) is a very potent activator of p32-kinase with an EC50 of 8 μM, and inhibits protein kinase C (PKC). D-erythro-Sphingosine (Erythrosphingosine) is also a PP2A activator.

Specifications

Chemical Name or Material D-erythro-Sphingosine
Physical Form Solid
CAS 123-78-4
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H37NO2
For Use With (Application) Cancer-Kinase/protease
Synonym Erythrosphingosine erythro-C18-Sphingosine trans-4-Sphingenine
Solubility Information DMSO : 50 mg/mL (166.95 mM; ultrasonic and warming and heat to 60°C)
SMILES CCCCCCCCCCCCC/C=C/[C@@H](O)[C@@H](N)CO
Molecular Weight (g/mol) 299.49
Formula Weight 299.49
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.