missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(5RS)-5-[[4-[2-(5-Ethylpyridin-2-yl)ethoxy]phenyl]methyl]-3-[2-(5-ethylpyridin-2-yl)ethyl]thiazolidine-2,4-dione, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
(5RS)-5-[[4-[2-(5-Ethylpyridin-2-yl)ethoxy]phenyl]methyl]-3-[2-(5-ethylpyridin-2-yl)ethyl]thiazolidine-2,4-dione
Specifications
Specifications
| Chemical Name or Material | (5RS)-5-[[4-[2-(5-Ethylpyridin-2-yl)ethoxy]phenyl]methyl]-3-[2-(5-ethylpyridin-2-yl)ethyl]thiazolidine-2,4-dione |
| CAS | 952188-00-0 |
| Analyte or Component Names | (5RS)-5-[[4-[2-(5-Ethylpyridin-2-yl)ethoxy]phenyl]methyl]-3-[2-(5-ethylpyridin-2-yl)ethyl]thiazolidine-2,4-dione |
| Shipping Condition | Room Temperature |
| Molecular Formula | C28 H31 N3 O3 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Pioglitazone Hydrochloride Imp. C (EP), Pioglitazone Imp. C (EP), (5RS)-5-[4-[2-(5-Ethylpyridin-2-yl)ethoxy]benzyl]-3-[2-(5-ethylpyridin-2-yl)ethyl]-1,3-thiazolidine-2,4-dione, Pioglitazone Hydrochloride Imp. C (Pharmeuropa), Pioglitazone Hydrochloride Impurity C, Pioglitazone Impurity C |
| InChI Formula | InChI=1S/C28H31N3O3S/c1-3-20-5-9-23(29-18-20)13-15-31-27(32)26(35-28(31)33)17-22-7-11-25(12-8-22)34-16-14-24-10-6-21(4-2)19-30-24/h5-12,18-19,26H,3-4,13-17H2,1-2H3 |
| SMILES | CCc1ccc(CCOc2ccc(CC3SC(=O)N(CCc4ccc(CC)cn4)C3=O)cc2)nc1 |
| IUPAC Name | 5-[[4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl]methyl]-3-[2-(5-ethylpyridin-2-yl)ethyl]-1,3-thiazolidine-2,4-dione |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction