missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5alpha-Epoxy-3-methoxy-17-methylmorphinan-6beta,14-diol Hydrochloride (6beta-Oxycodol Hydrochloride), Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
4,5alpha-Epoxy-3-methoxy-17-methylmorphinan-6beta,14-diol Hydrochloride (6beta-Oxycodol Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | 4,5alpha-Epoxy-3-methoxy-17-methylmorphinan-6beta,14-diol Hydrochloride (6beta-Oxycodol Hydrochloride) |
| Analyte or Component Names | 4,5alpha-Epoxy-3-methoxy-17-methylmorphinan-6beta,14-diol Hydrochloride (6beta-Oxycodol Hydrochloride) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H23 N O4 . H Cl |
| Certifications/Compliance | ISO 9001 |
| Synonym | Sildenafil Dimer, 4,5alpha-Epoxy-3-methoxy-17-methylmorphinan-6beta,14-diol Hydrochloride, 6beta-Oxycodol Hydrochloride |
| InChI Formula | InChI=1S/C18H23NO4.ClH/c1-19-8-7-17-14-10-3-4-12(22-2)15(14)23-16(17)11(20)5-6-18(17,21)13(19)9-10;/h3-4,11,13,16,20-21H,5-9H2,1-2H3;1H/t11-,13-,16+,17+,18-;/m1./s1 |
| SMILES | Cl.COc1ccc2C[C@H]3N(C)CC[C@@]45[C@@H](Oc1c24)[C@H](O)CC[C@@]35O |
| IUPAC Name | (4R,4aS,7R,7aR,12bS)-9-methoxy-3-methyl-1,2,4,5,6,7,7a,13-octahydro-4,12-methanobenzofuro[3,2-e]isoquinoline-4a,7-diol;hydrochloride |
| Molecular Weight (g/mol) | 353.84 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction