missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-(Disulfane-1,2-diyl)bis(N,N-diethylethanamine) Dihydrochloride, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
2,2'-(Disulfane-1,2-diyl)bis(N,N-diethylethanamine) Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | 2,2'-(Disulfane-1,2-diyl)bis(N,N-diethylethanamine) Dihydrochloride |
| CAS | 22194-38-3 |
| Analyte or Component Names | 2,2'-(Disulfane-1,2-diyl)bis(N,N-diethylethanamine) Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H28 N2 S2 . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Tiamulin Hydrogen Fumarate Imp. C (EP), Tiamulin Imp. C (EP), 2,2'-(Disulfane-1,2-diyl)bis(N,N-diethylethanamine) Dihydrochloride, Tiamulin Hydrogen Fumarate Imp. C (EP) as Dihydrochloride, Tiamulin Hydrogen Fumarate Impurity C as Dihydrochloride, Tiamulin Impurity C as Dihydrochloride |
| InChI Formula | InChI=1S/C12H28N2S2.2ClH/c1-5-13(6-2)9-11-15-16-12-10-14(7-3)8-4;;/h5-12H2,1-4H3;2*1H |
| SMILES | Cl.Cl.CCN(CC)CCSSCCN(CC)CC |
| IUPAC Name | 2-[2-(diethylamino)ethyldisulfanyl]-N,N-diethylethanamine;dihydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction