missing translation for 'onlineSavingsMsg'
Learn More

11,17-Dihydroxy-3-oxo-pregn-4-ene-21-carboxylic Acid γ-Lactone, TRC

Product Code. 30006356
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006356 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30006356 Supplier Toronto Research Chemicals Supplier No. TRCD88352050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 41850-19-5
Molecular Formula C22 H30 O4
Quantity 50 mg
Synonym (2'R,8S,9S,10R,11R,13S,14S)-11-Hydroxy-10,13-dimethyl-1,6,7,8,9,10,11,12,13,14,15,16-dodecahydro-3'H-spiro[cyclopenta[a]phenanthrene-17,2'-furan]-3,5'(2H,4'H)-dione
InChI Formula InChI=1S/C22H30O4/c1-20-8-5-14(23)11-13(20)3-4-15-16-6-9-22(10-7-18(25)26-22)21(16,2)12-17(24)19(15)20/h11,15-17,19,24H,3-10,12H2,1-2H3/t15-,16-,17+,19?,20-,21?,22+/m0/s1
SMILES C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@@]5(CCC(=O)O5)C4(C)C[C@@H](O)C23
IUPAC Name (8S,10R,11R,14S,17R)-11-hydroxy-10,13-dimethylspiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,5'-oxolane]-2',3-dione
Molecular Weight (g/mol) 358.471
Formula Weight 358.214
Chemical Name or Material 11,17-Dihydroxy-3-oxo-pregn-4-ene-21-carboxylic Acid Gamma-Lactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.