missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-4-(2,3,4-trimethoxybenzyl)piperazine Dihydrochloride (N-Methyltrimetazidine Dihydrochloride), Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
1-Methyl-4-(2,3,4-trimethoxybenzyl)piperazine Dihydrochloride (N-Methyltrimetazidine Dihydrochloride)
Specifications
Specifications
| Chemical Name or Material | 1-Methyl-4-(2,3,4-trimethoxybenzyl)piperazine Dihydrochloride (N-Methyltrimetazidine Dihydrochloride) |
| CAS | 53960-20-6 |
| Analyte or Component Names | 1-Methyl-4-(2,3,4-trimethoxybenzyl)piperazine Dihydrochloride (N-Methyltrimetazidine Dihydrochloride) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H24 N2 O3 . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Trimetazidine Dihydrochloride Imp. I (EP), Trimetazidine Imp. I (EP), 1-Methyl-4-(2,3,4-trimethoxybenzyl)piperazine Dihydrochloride, Trimetazidine Dihydrochloride Imp. I (EP) as Dihydrochloride, N-Methyltrimetazidine Dihydrochloride, Trimetazidine Dihydrochloride Impurity I as Dihydrochloride, Trimetazidine Impurity I as Dihydrochloride |
| InChI Formula | InChI=1S/C15H24N2O3.2ClH/c1-16-7-9-17(10-8-16)11-12-5-6-13(18-2)15(20-4)14(12)19-3;;/h5-6H,7-11H2,1-4H3;2*1H |
| SMILES | Cl.Cl.COc1ccc(CN2CCN(C)CC2)c(OC)c1OC |
| IUPAC Name | 1-methyl-4-[(2,3,4-trimethoxyphenyl)methyl]piperazine;dihydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction