missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc, ILZRO 16, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Zinc, ILZRO 16, 50g
Specifications
Specifications
| Chemical Name or Material | Cadmium/Manganese/Magnesium/Nickel/Lead/Iron/Aluminium/Chromium/Titanium/Copper/Tin |
| CAS | 7440-66-6 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Cadmium 0.0005 %/Manganese 0.0013 %/Magnesium 0.0029 %/Nickel 0.0038 %/Lead 0.0042 %/Iron 0.011 %/Aluminium 0.029 %/Chromium 0.083 %/Titanium 0.278 %/Copper 0.79 %/Tin (0.0007) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction