missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead, Alloy, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Lead, Alloy, 50g
Specifications
Specifications
| Chemical Name or Material | Cadmium/Nickel/Tellurium/Silver/Tin/Copper/Bismuth/Arsenic/Antimony |
| CAS | 7439-92-1 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Cadmium 0.00024 %/Nickel 0.0005 %/Tellurium 0.0006 %/Silver 0.0016 %/Tin 0.013 %/Copper 0.0245 %/Bismuth 0.0602 %/Arsenic 0.683 %/Antimony 1.54 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction