missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroacetaldehyde Hydrate, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | Chloralhydrate |
| CAS | 302-17-0 |
| Molecular Formula | C2 H3 Cl3 O2 |
| Quantity | 50 g |
| Synonym | Chloral hydrate (8CI), 2,2,2-Trichloro-1,1-ethanediol, 1,1,1-Trichloro-2,2-dihydroxyethane, Bi 3411, Chloral','monohydrate, Chloraldurat, Dormal, Escre, Fagron, Hydral, Lorinal, NSC 3210, Noctec, Nycoton, Nycton, Phaldrone, Rectules, SK Chloral hydrate, Somnos, Trawotox, Trichloroacetaldehyde hydrate, Trichloroacetaldehyde monohydrate |
| InChI Formula | InChI=1S/C2H3Cl3O2/c3-2(4,5)1(6)7/h1,6-7H |
| SMILES | OC(O)C(Cl)(Cl)Cl |
| IUPAC Name | 2,2,2-trichloroethane-1,1-diol |
| Molecular Weight (g/mol) | 165.4 |
| Formula Weight | 163.9199 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction