missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solder, Lead-Free, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Solder, Lead-Free, 50g
Specifications
Specifications
| Chemical Name or Material | Aluminium/Zinc/Cadmium/Gold/Nickel/Germanium/Iron/Phosphorus/Indium/Arsenic/Bismuth/Lead/Antimony/Copper/Silver |
| CAS | 7440-31-5 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Aluminium 0.0007 %/Zinc 0.001 %/Cadmium 0.0015 %/Gold 0.0025 %/Nickel 0.0039 %/Germanium 0.0049 %/Iron 0.0085 %/Phosphorus 0.011 %/Indium 0.0165 %/Arsenic 0.0173 %/Bismuth 0.0364 %/Lead 0.038 %/Antimony 0.076 %/Copper 0.097 %/Silver 1.002 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction