missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Cr/Mo/Co, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Nickel, Ni/Cr/Mo/Co, 50g
Specifications
Specifications
| Chemical Name or Material | Aluminium/Titanium/Copper/Iron/Manganese/Cobalt/Molybdenum/Chromium |
| CAS | 7440-02-0 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Aluminium 0.02 %/Titanium 0.02 %/Copper 0.09 %/Iron 0.4 %/Manganese 0.4 %/Cobalt 10.5 %/Molybdenum 10.7 %/Chromium 21.8 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction