missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium, Mg/Al/Rare Earth, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Magnesium, Mg/Al/Rare Earth, 50g
Specifications
Specifications
| Chemical Name or Material | Beryllium/Nickel/Copper/Iron/Gadolinium/Zinc/Silicon/Praseodymium/Neodymium/Lanthanum/Manganese/Cerium/Aluminium |
| CAS | 7439-95-4 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Beryllium 0.0007 %/Nickel 0.0016 %/Copper 0.0017 %/Iron 0.0024 %/Gadolinium 0.038 %/Zinc 0.05 %/Silicon 0.068 %/Praseodymium 0.069 %/Neodymium 0.175 %/Lanthanum 0.374 %/Manganese 0.516 %/Cerium 0.83 %/Aluminium 4.56 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction