missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Aluminum Bronze, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Copper, Aluminum Bronze, 50g
Specifications
Specifications
| Chemical Name or Material | Silver/Magnesium/Arsenic/Silicon/Lead/Tin/Zinc/Cobalt/Manganese/Iron/Nickel/Aluminium/Copper/Phosphorus |
| CAS | 7440-50-8 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Silver 0.0082 %/Magnesium 0.0104 %/Arsenic 0.0116 %/Silicon 0.073 %/Lead 0.0749 %/Tin 0.12 %/Zinc 0.126 %/Cobalt 0.139 %/Manganese 0.787 %/Iron 2.71 %/Nickel 5.42 %/Aluminium 9.69 %/Copper 80.77 %/Phosphorus (0.006) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction