missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 5 12-14 kD MWCO Standard RC Dry Dialysis Kits
Click to view available options
Diameter (Metric):
48 mm
89 mm
Length (Metric):
18 mL/cm vol./length
62 mL/cm vol./length
Quantity:
1 m Tubing/Kit
5 m Tubing/Kit
Unit Size:
1m
5m
Description
Includes
5 m of Spectra/Por 5 dialysis tubing and 2 closures
- For reinforced for weight applications, or evaluating dialysis tubing for dialysis application
- Packaged dry without glycerin (no soaking required)
Specifications
Specifications
| MWCO | 12 to 14 kDa |
| Diameter (Metric) | 89 mm |
| Length (Metric) | 62 mL/cm vol./length |
| Quantity | 5 m Tubing/Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction