missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Spectra/Por™ 2 12-14 kD MWCO Standard RC Dry Dialysis Kits

Product Code. 11541040
Click to view available options
Diameter (Metric):
6.5 mm
16 mm
29 mm
67 mm
76 mm
Length (Metric):
0.32 mL/cm vol./length
2 mL/cm
34 mL/cm vol./length
46 mL/cm vol./length
5 m
6.4 mL/cm
Quantity:
1 m Tubing/Kit
5 m Tubing/Kit
Each
Unit Size:
1m
5m
This item is not returnable. View return policy

Product Code. 11541040

Brand: Spectrum™ 132680T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • 1m or 5m of Spectra/Por 2 dialysis tubing
  • 1 Standard Spectra/Por Closure (for buoyance)
  • 1 weighted Spectra/Por Closure (for orientation)
  • 2 adhesive tubing openers

With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated

  • For higher permeability applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry with glycerin (soak in water)

Specifications

Product Type Trial Kit
MWCO 12 to 14 kDa
Material Regenerated Cellulose
Diameter (Metric) 29 mm
Diameter (English) 1.1 in.
Length (English) 16 ft.
Length (Metric) 5 m
Capacity (Metric) 6.4 mL
Kit Contents 5m roll of Spectra/Por™ 1, 2 or 3 dialysis tubing, 1 standard spectra/por closure (aids buoyancy), 1 weighted spectra/por closure (aids vertical orientation), 5 opening picks
Quantity Each
For Use With (Application) Ideal for small volume dialysis or evaluation of membrane for an application
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.