missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Spectra/Por™ 1 6-8 kD MWCO Standard RC Dry Dialysis Kits

Product Code. 11521040
Change view
Click to view available options
Diameter (Metric):
6.4 mm
14.6 mm
20.4 mm
25.5 mm
32 mm
64 mm
76 mm
Length (Metric):
0.32 mL/cm vol./length
1.7 mL/cm
3.3 mL/cm vol./length
32 mL/cm vol./length
46 mL/cm vol./length
5 m
5.1 mL/cm vol./length
7.9 mL/cm vol./length
Quantity:
1 m Tubing/Kit
5 m Tubing/Kit
Each
Unit Size:
1m
5m
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Length (Metric) Quantity Unit Size
11521040 32 mm 5 m Each 5m
15370752 76 mm 46 mL/cm vol./length 5 m Tubing/Kit 5m
15350752 64 mm 32 mL/cm vol./length 5 m Tubing/Kit 5m
15380752 76 mm 46 mL/cm vol./length 1 m Tubing/Kit 1m
15330752 32 mm 7.9 mL/cm vol./length 1 m Tubing/Kit 1m
15360752 64 mm 32 mL/cm vol./length 1 m Tubing/Kit 1m
15320752 25.5 mm 5.1 mL/cm vol./length 1 m Tubing/Kit 1m
15300752 20.4 mm 3.3 mL/cm vol./length 1 m Tubing/Kit 1m
15310752 25.5 mm 5.1 mL/cm vol./length 5 m Tubing/Kit 5m
15380742 14.6 mm 1.7 mL/cm 1 m Tubing/Kit 1m
15370742 14.6 mm 1.7 mL/cm 5 m Tubing/Kit 5m
15350742 6.4 mm 0.32 mL/cm vol./length 5 m Tubing/Kit 5m
15360742 6.4 mm 0.32 mL/cm vol./length 1 m Tubing/Kit 1m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Product Code. 11521040 Supplier Spectrum™ Supplier No. 132665T
  • ONLINE EXCLUSIVE PRICE
    £170.61
    £145.96 / 5m
    Save £24.65
    14% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • 1m or 5m of Spectra/Por 1 dialysis tubing
  • 1 Standard Spectra/Por Closure (for buoyance)
  • 1 weighted Spectra/Por Closure (for orientation)
  • 2 adhesive tubing openers

With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated

  • For basic dialysis applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry with glycerin (soak in water)

Specifications

Product Type Trial Kit
MWCO 6 to 8 kDa
Membrane Regenerated Cellulose
Diameter (Metric) 32 mm
Diameter (English) 1.2 in.
Length (English) 16 ft.
Length (Metric) 5 m
Capacity (Metric) 7.9 mL
Kit Contents 5m roll of Spectra/Por™ 1, 2 or 3 dialysis tubing, 1 standard spectra/por closure (aids buoyancy), 1 weighted spectra/por closure (aids vertical orientation), 5 opening picks
Quantity Each
For Use With (Application) Ideal for small volume dialysis or evaluation of membrane for an application
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.