missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs 100 kD Biotech CE Dialysis Kits

Product Code. 15320682
Change view
Click to view available options
Diameter (Metric):
6.4 mm
10 mm
15 mm
20 mm
Length (Metric):
0.32 mL/cm vol./length
0.79 mL/cm vol./length
1.8 mL/cm vol./length
3.1 mL/cm vol./length
Unit Size:
1m
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Length (Metric) Unit Size
15320682 20 mm 3.1 mL/cm vol./length 1m
15310682 15 mm 1.8 mL/cm vol./length 1m
15300682 6.4 mm 0.32 mL/cm vol./length 1m
11500980 10 mm 0.79 mL/cm vol./length Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15320682 Supplier Spectrum™ Supplier No. 131420T
  • ONLINE EXCLUSIVE PRICE
    £362.02
    £164.09 / 1m
    Save £197.93
    54% Off
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

  • 1m Biotech CE Tubing in 0.05% sodium azide
  • Two universal tubing closures
  • Five opening picks

With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane

Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC

  • Hydrophilic, symmetric porositiy
  • For small tubing demand applications or evaluating a specific dialysis membrane
  • Includes 1m tubing with pair of tubing closures and 5 opening picks

Specifications

MWCO 100 kDa
Diameter (Metric) 20 mm
Length (Metric) 3.1 mL/cm vol./length
Quantity 1 m Tubing/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.