Learn More
Spectrum™ Labs 100 kD Biotech CE Dialysis Kits
With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane
Brand: Spectrum™ 131408T
Code : NEW
Additional Details : Weight : 0.10000kg
Includes:
- 1m Biotech CE Tubing in 0.05% sodium azide
- Two universal tubing closures
- Five opening picks
Description
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
- Hydrophilic, symmetric porositiy
- For small tubing demand applications or evaluating a specific dialysis membrane
- Includes 1m tubing with pair of tubing closures and 5 opening picks
Specifications
100 kDa | |
6.4 mm | |
0.32 mL/cm vol./length | |
1 m Tubing/Kit |