missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs 3.5 - 5 kD Biotech CE Dialysis Kits

With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane

Brand:  Spectrum™ 131192T

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15380662

  • £136.00 / 1m
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes:
  • 1m Biotech CE Tubing in 0.05% sodium azide
  • Two universal tubing closures
  • Five opening picks

Description

Description

Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC

  • Hydrophilic, symmetric porositiy
  • For small tubing demand applications or evaluating a specific dialysis membrane
  • Includes 1m tubing with pair of tubing closures and 5 opening picks

Specifications

Specifications

3.5 to 5 kDa
6.4 mm
0.32 mL/cm vol./length
1 m Tubing/Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers