missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 7 25,000 D MWCO Standard RC Pre-Treated Dialysis Kits
With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds
£144.49 - £165.00
Specifications
MWCO | 25 kDa |
---|---|
Quantity | 1 m Tubing/Kit |
Includes:
- 1meter of pre-treated tubing, a pair of closures and 5 opening picks
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
15310712
|
Spectrum™
132127T |
3.7 mL/cm vol./length | 22 mm |
£146.00
1m |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15300712
|
Spectrum™
132126T |
2.5 mL/cm vol./length | 18 mm |
£165.00
1m |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15390702
|
Spectrum™
132 125T |
0.45 mL/cm vol./length | 7.5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Hydrophilic, symmetric porositiy
- Equilibrated in water
- Packaged wet in preservative solution of 0.05% sodium azide
- For small experiments or qualifying a membrane for a particular application
Specifications
25 kDa | |
1 m Tubing/Kit |