missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBind™ Flex Western Device

Product Code. 17134609
Change view
Click to view available options
Type:
Western Device
Quantity:
1 device
2 devices
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Quantity Unit Size
17134609 Western Device 2 devices Pack of 2
15357268 Western Device 1 device Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17134609 Supplier Invitrogen™ Supplier No. SLF20002PK
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Allows better antigen-antibody interaction and solutions flow between the iBind™ Card and membrane for maximum interaction between protein and antibody

The iBind™ Flex Western System offers automated western blot processing and can accommodate various blot formats including midi, mini, and vertically cut strips. Based on Sequential Lateral Flow (SLF) technology, all blocking, antibody incubation and washing steps during immunodetection are hands-free. Compared to manual methods, the iBind Flex Western System requires less primary antibody, enables reproducible results and is compatible with downstream detection protocols.

The Invitrogen™ iBind™ Flex Western System offers:
  • Flexible options—optimize antibody use, change formats and reduce hands-on time
  • Cost savings—use up to 80% less primary antibody
  • Reproducibility—automated processing provides improved blot-to-blot consistency
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Capacity 1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
Detection Method Fluorescence, Chemiluminescence, Chromogenic
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Product Line iBind Flex
Running Time ∼2.5 hr.
Throughput 1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
Type Western Device
Quantity 2 devices
Running Dimension Horizontal
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.