missing translation for 'onlineSavingsMsg'
Learn More

R&DSystems™ Recombinant Human KGF/FGF-7 Protein, CF

Product Code. 30019017
Click to view available options
Quantity:
10 μg
100 μg
50 μg
500 μg
Unit Size:
100µg
10µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 30019017

Brand: R&DSystems™ 11537KG500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CHO expressed

Measured in a cell proliferation assay using Ba/F3 mouse pro B cells transfected with human FGF RIIb. The ED50 for this effect is 4.00-40.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P21781.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, Na2SO4 and EDTA with Trehalose.
Gene ID (Entrez) 2252
Molecular Weight (g/mol) M.W.-observed: 15-18 kDa, under reducing conditions, M.W.-theroretical: 16 kDa
Quantity 500 μg
Source Chinese Hamster Ovary cell line, CHO-derived human KGF/FGF-7 protein Ser55-Thr194
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias FGF7, FGF-7, fibroblast growth factor 7HBGF-7, HBGF7, HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGFfibroblast growth factor 7 (keratinocyte growth factor)
Conjugate Unconjugated
Reconstitution Reconstitute 10 μg size at 100 μg/mL and the other sizes at 200 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein KGF/FGF-7
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.