missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL8/MCP-2 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB790-500
This item is not returnable.
View return policy
Description
CCL8/MCP-2 Monoclonal specifically detects CCL8/MCP-2 in Mouse samples. It is validated for Western Blot.
Specifications
| CCL8/MCP-2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C motif chemokine 8, chemokine (C-C motif) ligand 8, HC14, MCP2, MCP-2, member 8 (monocyte chemotactic protein 2), monocyte chemoattractant protein 2, SCYA10, small-inducible cytokine A8 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6355 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 146123 | |
| Western Blot 1 ug/mL | |
| Q9Z121 | |
| CCL8 | |
| E. coli-derived recombinant mouse CCL8/MCP-2 Glu20-Pro97 Accession # Q9Z121 | |
| 500 μg | |
| Primary | |
| Detects mouse CCL8/MCP-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant cotton rat CCL3, 4, 5, recombinant human CCL1, 2, 3, 4, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, 27, 28, recombinant mouse CCL1, 3, 4, 5, 6, 7, 9/10MIP-1 gamma , 11, 12, 17, 19, 20, 21, 22, 24, 25, 27, or recombinant rat CCL20 is observed. | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction