missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tuning Solution for PerkinElmer Instruments: 7Li, Be, Mg, Co, Y, In, Ba, Ce, Tl, Pb, U @ 10 μg/L in 2% HNO3, VHG™
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
Tuning Solution for PerkinElmer Instruments: 7Li, Be, Mg, Co, Y, In, Ba, Ce, Tl, Pb, U @ 10 μg/L in 2% HNO3
Specifications
Specifications
| Chemical Name or Material | Barium/Uranium/Beryllium/Cerium/Cobalt/Lead/Thallium/Yttrium/Indium/Lithium/Magnesium |
| CAS | 7440-39-3, 7440-61-1, 7440-41-7, 7440-45-1, 7440-48-4, 7439-92-1 |
| Methods | ICP-MS |
| Analyte or Component Names | Barium/Uranium/Beryllium/Cerium/Cobalt/Lead/Thallium/Yttrium/Indium/Lithium/Magnesium |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | Barium 10 μg/L/Uranium 10 μg/L/Beryllium 10.01 μg/L/Cerium 10.01 μg/L/Cobalt 10.01 μg/L/Lead 10.01 μg/L/Thallium 10.01 μg/L/Yttrium 10.01 μg/L/Indium 10.02 μg/L/Lithium 10.02 μg/L/Magnesium 10.02 μg/L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction