missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchlorate Standard: ClO4- @ 1000 μg/mL in H2O, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
Perchlorate Standard: ClO4- @ 1000 μg/mL in H2O
Specifications
Specifications
| Chemical Name or Material | Perchlorate |
| CAS | 14797-73-0 |
| Methods | Ion Chromatography/Wet Chemistry |
| Analyte or Component Names | Perchlorate |
| Shipping Condition | Room Temperature |
| Molecular Formula | (Cl O4)- |
| Synonym | ClO4− |
| IUPAC Name | Perchlorate |
| Solvent or Matrix | Water |
| Molecular Weight (g/mol) | 99.45 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction