missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalate Standard: C2O4(-2) @ 10000 μg/mL in H2O, VHG™
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
Oxalate Standard: C2O4(-2) @ 10000 μg/mL in H2O
Specifications
Specifications
| Chemical Name or Material | Oxalate |
| CAS | 338-70-5 |
| Methods | Ion Chromatography |
| Analyte or Component Names | Oxalate |
| Molecular Formula | C2O42− |
| Synonym | (C2 O4)2- |
| IUPAC Name | Oxalate |
| Solvent or Matrix | Water |
| Molecular Weight (g/mol) | 88.02 |
| Quantity | 500 mL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction