missing translation for 'onlineSavingsMsg'
Learn More

ICS Interference A: Fe @ 2000; Al, Ca, Mg @ 5000 μg/mL in 20% HCl, VHG™

Product Code. 30002995
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
This item is not returnable. View return policy

Product Code. 30002995

Brand: VHG Labs VHGICSA500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ICS Interference A: Fe @ 2000; Al, Ca, Mg @ 5000 μg/mL in 20% HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iron/Aluminium/Calcium/Magnesium
CAS 7439-89-6, 7429-90-5, 7440-70-2, 7439-95-4
Methods ICP
Analyte or Component Names Iron/Aluminium/Calcium/Magnesium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HCl
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Iron 2000 μg/mL/Aluminium 5000 μg/mL/Calcium 5000 μg/mL/Magnesium 5000 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.