missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elements in Engine Coolant, QC Standard: Na @ 5000; K @ 2500; P @ 1000; B @ 250; Ca @ 50; Fe, Pb @ 25; Zn @ 20; Mg @ 10; Al, Cu @ 5 ug/g in 3% HNO3, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
Elements in Engine Coolant, QC Standard: Na @ 5000; K @ 2500; P @ 1000; B @ 250; Ca @ 50; Fe, Pb @ 25; Zn @ 20; Mg @ 10; Al, Cu @ 5 ug/g in 3% HNO3
Specifications
Specifications
| Chemical Name or Material | Aluminium/Copper/Magnesium/Zinc/Iron/Lead/Calcium/Boron/Phosphorus/Potassium/Sodium |
| CAS | 7429-90-5, 7440-50-8, 7439-95-4, 7440-66-6, 7439-89-6, 7439-92-1 |
| Methods | ICP |
| Analyte or Component Names | Aluminium/Copper/Magnesium/Zinc/Iron/Lead/Calcium/Boron/Phosphorus/Potassium/Sodium |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | Aluminium 5 μg/g/Copper 5 μg/g/Magnesium 10 μg/g/Zinc 20 μg/g/Iron 25 μg/g/Lead 25 μg/g/Calcium 50 μg/g/Boron 250 μg/g/Phosphorus 1000 μg/g/Potassium 2500 μg/g/Sodium 5000 μg/g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction