missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Common Elements Mix 1 Standard: Cd, Co, Cr, Cu, Fe, Mn, Ni, V, Zn @ 100 μg/mL in 5% HNO3, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
Common Elements Mix 1 Standard: Cd, Co, Cr, Cu, Fe, Mn, Ni, V, Zn @ 100 μg/mL in 5% HNO3
Specifications
Specifications
| Chemical Name or Material | Cadmium/Chromium/Cobalt/Copper/Iron/Nickel/Vanadium/Zinc/Manganese |
| CAS | 7440-43-9, 7440-47-3, 7440-48-4, 7440-50-8, 7439-89-6, 7440-02-0 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Cadmium/Chromium/Cobalt/Copper/Iron/Nickel/Vanadium/Zinc/Manganese |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | Cadmium 100 μg/mL/Chromium 100 μg/mL/Cobalt 100 μg/mL/Copper 100 μg/mL/Iron 100 μg/mL/Nickel 100 μg/mL/Vanadium 100 μg/mL/Zinc 100 μg/mL/Manganese 100.1 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction