Learn More
500ML Buffer Standard pH 9.225 20oC ISO 17034 CRMCertified Reference Material NIST Value
Brand:  Reagecon CRM109225
500ML Buffer Standard pH 9.225 20oC ISO 17034 CRMCertified Reference Material NIST Value
Additional Details : CAS Number : 1303-96-4 Weight : 0.52000kg
Product Code. 17973802
  • £35.85 / 500mL
Estimated Shipment: 25-06-2024