missing translation for 'onlineSavingsMsg'
Learn More

Racanisodamine, MedChemExpress

Product Code. 16452396
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452396 500 mg 500mg
16449176 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16452396 Supplier Medchem Express Supplier No. HYN2064

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Racanisodamine is one of the racemic isomers of anisodamine, resembles anisodamine in pharmacological effect. Racanisodamine is a non-selective muscarinic antagonist, used as a component of eye drops for myopic control.

Racanisodamine is one of the racemic isomers of anisodamine, resembles anisodamine in pharmacological effect. Racanisodamine is a non-selective muscarinic antagonist, used as a component of eye drops for myopic control.

Specifications

Chemical Name or Material Racanisodamine
CAS 17659-49-3
Color White
Purity Grade Notes Research
Molecular Formula C17H23NO4
Quantity 500 mg
Solubility Information DMSO : ≥ 125 mg/mL (409.34 mM)
SMILES O=C(OC1CC(N2C)CC(O)C2C1)C(CO)C3=CC=CC=C3
Molecular Weight (g/mol) 305.37
Formula Weight 305.37
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.