missing translation for 'onlineSavingsMsg'
Learn More

rac 3,4-Dihydroxymandelic Acid, TRC

Product Code. 30011277
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011277 500 mg 500mg
30133025 5 g 5g
30008404 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011277 Supplier Toronto Research Chemicals Supplier No. TRCD453000500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 775-01-9
Molecular Formula C8 H8 O5
Molecular Weight (g/mol) 184.15
IUPAC Name 2-(3,4-dihydroxyphenyl)-2-hydroxyacetic acid
SMILES OC(C(=O)O)c1ccc(O)c(O)c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H8O5/c9-5-2-1-4(3-6(5)10)7(11)8(12)13/h1-3,7,9-11H,(H,12,13)
Formula Weight 184.0372
Percent Purity >95%
Chemical Name or Material (±)-3,4-Dihydroxymandelic Acid (DOMA)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.