missing translation for 'onlineSavingsMsg'
Learn More

LY294002, MedChemExpress

Product Code. 16461526
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16461526

Brand: Medchem Express HY10108

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LY294002 is a broad-spectrum inhibitor of PI3K with IC50s of 0.5, 0.57, and 0.97 μM for PI3Kα, PI3Kδ and PI3Kβ, respectively. LY294002 also inhibits CK2 with an IC50 of 98 nM. LY294002 is a competitive DNA-PK inhibitor that binds reversibly to the kinase domain of DNA-PK with an IC50 of 1.4 μM. LY294002 is an apoptosis activator.

LY294002 is a broad-spectrum inhibitor of PI3K with IC50s of 0.5, 0.57, and 0.97 μM for PI3Kα, PI3Kδ and PI3Kβ, respectively. LY294002 also inhibits CK2 with an IC50 of 98 nM. LY294002 is a competitive DNA-PK inhibitor that binds reversibly to the kinase domain of DNA-PK with an IC50 of 1.4 μM. LY294002 is an apoptosis activator.

Specifications

Chemical Name or Material LY294002
CAS 154447-36-6
Color White
Purity Grade Notes Research
Molecular Formula C19H17NO3
Quantity 500 mg
Solubility Information DMSO : 50 mg/mL (162.69 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (162.69 mM; Need ultrasonic)
SMILES O=C1C=C(OC2=C1C=CC=C2C3=CC=CC=C3)N4CCOCC4
Molecular Weight (g/mol) 307.34
Formula Weight 307.34
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.