missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Urethane, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Diethyl Urethane |
| CAS | 3553-80-8 |
| Molecular Formula | C7H15NO2 |
| Quantity | 500 mg |
| Synonym | Ethyl N,N-diethylcarbamate |
| InChI Formula | InChI=1S/C7H15NO2/c1-4-8(5-2)7(9)10-6-3/h4-6H2,1-3H3 |
| SMILES | O=C(OCC)N(CC)CC |
| IUPAC Name | ethyl diethylcarbamate |
| Molecular Weight (g/mol) | 145.2 |
| Formula Weight | 145.11 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction