missing translation for 'onlineSavingsMsg'
Learn More

Cisplatin, MedChemExpress

Product Code. 16441846
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16441846

Brand: Medchem Express HY17394

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cisplatin (CDDP) is an antineoplastic chemotherapy agent by cross-linking with DNA and causing DNA damage in cancer cells. Cisplatin activates ferroptosis and induces autophagy.

Cisplatin (CDDP) is an antineoplastic chemotherapy agent by cross-linking with DNA and causing DNA damage in cancer cells. Cisplatin activates ferroptosis and induces autophagy.

Specifications

Chemical Name or Material Cisplatin
Physical Form Powder
CAS 15663-27-1
Color Yellow
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula Cl2H6N2Pt
For Use With (Application) Cancer-programmed cell death
Synonym cis-Platinum CDDP cis-Diaminodichloroplatinum
Solubility Information DMSO : 250 mg/mL (833.19 mM; Need ultrasonic; DMSO can inactivate Cisplatin's activity) ∣DMF : 5 mg/mL (16.66 mM; Need ultrasonic; DMSO can inactivate Cisplatin's activity) ∣H2O : 1 mg/mL (3.33 mM; Need ultrasonic; DMSO can inactivate Cisplatin's activity)
SMILES Cl[Pt](Cl)([NH3])[NH3]
Molecular Weight (g/mol) 300.05
Formula Weight 300.05
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.