Learn More
Melford Miller's LB Agar, 1G Capsules
Description
Super convenient.
Super convenient. Eliminate messy weighing. Consistant and accurate measurement. Convenient premeasured and pre-mixed capsules. Prebuffered media eliminates pH adjustments.
Simply add 40 capsules to one liter of water, sterilize by autoclaving for 15 minutes at 121°C. Capsules contain all necessary ingredients for maintenance and propagation of E. Coli.
Ingredients:
Casein Digest Peptone: 10g/L
Yeast Extract: 5g/L
Sodium Chloride: 10g/L
Agar:15g/L
TRIS/TRIS HCl: 1.5g/L
Specifications
Specifications
| Quantity | 500 g |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.