missing translation for 'onlineSavingsMsg'
Learn More

Melford LB Broth, Powder

Product Code. 30018415
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 30018415

Brand: Melford L24060500.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For propagation and maintenance of E.

For propagation and maintenance of E. Coli. Low Salt Formula. Molecular Biology Grade.

Dissolve 20 grams in 1 liter of distilled water. Then adjust pH to 7.2. Heat and frequently agitate to completely dissolve. Sterilize by autoclaving for 15 minutes.

Ingredients:

Peptone: 10 g/L

Yeast Extract: 5 g/L

Sodium Chloride: 5 g/L

Specifications

Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.