missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Spectra/Por™ 7 3500 D MWCO Standard RC Pre-Treated Dialysis Kits

With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds

Brand:  Spectrum™ 132112T

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15360692

  • £165.00 / 1m
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes:
  • 1meter of pre-treated tubing, a pair of closures and 5 opening picks

Description

Description

  • Hydrophilic, symmetric porositiy
  • Equilibrated in water
  • Packaged wet in preservative solution of 0.05% sodium azide
  • For small experiments or qualifying a membrane for a particular application

Specifications

Specifications

3.5 kDa
34 mm
9.3 mL/cm vol./length
1 m Tubing/Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers