missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human NRG1/HRG1-alpha EGF domain Fc Protein
Click to view available options
Quantity:
50 μg
500 μg
Unit Size:
500µg
50µg
Description
Measured in a serum-free cell proliferation assay using MCF-7 human breast cancer cells. Karey, K.P. et al. (1988) Cancer Research 48:4083. The ED50 for this effect is 4.00-40.0 ng/mL.
Specifications
Specifications
| Accession Number | NP_039254.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 3084 |
| Molecular Weight (g/mol) | M.W. (Observed): 36-43 kDa, under reducing conditions., M.W. (Theoretical): 32 kDa |
| Quantity | 500 μg |
| Source | Human embryonic kidney cell, HEK293-derived human Neuregulin-1/NRG1 protein Human NRG1/HRG1-alpha EGF domain (Ser177-Pro226) (N-terminus) Accession # NP_039254.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Gene Alias | ARIA, GGF2, GGFglial growth factor, Heregulin-1, HGL, HGLneu differentiation factor, HRG, HRG1, HRG1-alpha, HRG1-beta 1, HRGAneuregulin 1 type IV beta 1a, MST131, MSTP131, NDFheregulin, alpha (45kD, ERBB2 p185-activator), neuregulin 1, neuregulin 1 type IV beta 3, Neuregulin1, NRG1, pro-neuregulin-1, membrane-bound isoform, pro-NRG1, sensory and motor neuron derived factor, SMDF |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction