Learn More
Thermo Scientific Eutech 500 ML Conductivity standard Eutech 111,800uS/cm+/-0.02pH (500mL)
Brand:  Thermo Scientific Eutech 01X211244
500 ML Conductivity standard Eutech 111,800uS/cm+/-0.02pH (500mL)
Product Code. 12848923